accessibility menu, dialog, popup

UserName

H3CNCH3O

CAS RN 1696-17-9

IUPAC Name: N,N-diethylbenzamide
Synonyms: N,N-diethylbenzamide
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: JLNGEXDJAQASHD-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results