missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzyl-4,13-diaza-18-crown 6-Ether 98.0+%, TCI America™

Catalog No. p-7127229
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23211G 1 g
D23215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23211G Supplier TCI America Supplier No. D23211G
Only null left

Chemical Identifiers

CAS 69703-25-9
Molecular Formula C26H38N2O4
Molecular Weight (g/mol) 442.6
MDL Number MFCD00010831
InChI Key WAHZGOBRKWVALN-UHFFFAOYSA-N
PubChem CID 3308751
IUPAC Name 7,16-dibenzyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
SMILES C1COCCOCCN(CCOCCOCCN1CC2=CC=CC=C2)CC3=CC=CC=C3

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 1 g
Synonym N,N′C-Dibenzyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
Formula Weight 442.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dibenzyl-4,13-diaza-18-crown 6-Ether