accessibility menu, dialog, popup

UserName

OONOON

CAS RN 69703-25-9

IUPAC Name: 7,16-dibenzyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
Synonyms: 7,16-dibenzyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
Molecular Weight (g/mol): 442.6
Molecular Formula: C26H38N2O4
InChi Key: WAHZGOBRKWVALN-UHFFFAOYSA-N
SMILES: C(N1CCOCCOCCN(CC2=CC=CC=C2)CCOCCOCC1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results