missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dibenzyl-2-aminoethanol 98.0+%, TCI America™

Catalog No. D0153500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0153500G 500 g
D015325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0153500G Supplier TCI America Supplier No. D0153500G
Only null left

Chemical Identifiers

CAS 101-06-4
Molecular Formula C16H19NO
Molecular Weight (g/mol) 241.334
MDL Number MFCD00020574
InChI Key WTTWSMJHJFNCQB-UHFFFAOYSA-N
Synonym N,N-Dibenzylethanolamine, N-(2-Hydroxyethyl)dibenzylamine
PubChem CID 22657
IUPAC Name 2-(dibenzylamino)ethanol
SMILES C1=CC=C(C=C1)CN(CCO)CC2=CC=CC=C2

Specifications

Color White-Yellow
Boiling Point 206°C
Quantity 500 g
Formula Weight 241.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material N,N-Dibenzyl-2-aminoethanol
EINECSNumber (3)-0314
RTECSNumber KJ8125000
TSCA Yes