accessibility menu, dialog, popup

UserName

CAS RN 101-06-4

HON

CAS RN 101-06-4

IUPAC Name: 2-(dibenzylamino)ethan-1-ol
Synonyms: 2-(dibenzylamino)ethanol
Molecular Weight (g/mol): 241.33
Molecular Formula: C16H19NO
InChi Key: WTTWSMJHJFNCQB-UHFFFAOYSA-N
SMILES: OCCN(CC1=CC=CC=C1)CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results