missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diacetyl-1,4-phenylenediamine 98.0+%, TCI America™

Catalog No. D00685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00685G 5 g
D006825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00685G Supplier TCI America Supplier No. D00685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 140-50-1
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.218
MDL Number MFCD00026142
InChI Key KVEDKKLZCJBVNP-UHFFFAOYSA-N
Synonym 1,4-diacetamidobenzene, n,n'-diacetyl-1,4-phenylenediamine, n,n'-p-phenylenebisacetamide, p-phenylenediacetamide, n,n'-diacetyl-p-phenylenediamine, 4'-acetamidoacetanilide, 1,4-bisacetamidobenzene, acetamide, n,n'-1,4-phenylenebis, n-4-acetamidophenyl acetamide, n,n'-p-phenylene di acetamide
PubChem CID 67324
IUPAC Name N-(4-acetamidophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)NC(=O)C

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 192.22
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diacetyl-1,4-phenylenediamine