missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Di(2-hydroxyethyl)glycine 99.0+%, TCI America™
$46.60 - $490.25
Chemical Identifiers
| CAS | 150-25-4 |
|---|---|
| Molecular Formula | C6H12NNaO4 |
| Molecular Weight (g/mol) | 185.16 |
| MDL Number | MFCD00004295 |
| InChI Key | MFBDBXAVPLFMNJ-UHFFFAOYSA-M |
| Synonym | bicine, n,n-bis 2-hydroxyethyl glycine, diethylolglycine, dihydroxyethylglycine, diethanol glycine, n,n-di 2-hydroxyethyl glycine, bicene, 2-bis 2-hydroxyethyl amino acetic acid, n,n-dihydroxyethyl glycine, n,n-dihydroxyethylglycine |
| PubChem CID | 8761 |
| ChEBI | CHEBI:40957 |
| IUPAC Name | sodium 2-[bis(2-hydroxyethyl)amino]acetate |
| SMILES | [Na+].OCCN(CCO)CC([O-])=O |
Chemical Identifiers
| 150-25-4 | |
| 185.16 | |
| MFBDBXAVPLFMNJ-UHFFFAOYSA-M | |
| 8761 | |
| sodium 2-[bis(2-hydroxyethyl)amino]acetate |
| C6H12NNaO4 | |
| MFCD00004295 | |
| bicine, n,n-bis 2-hydroxyethyl glycine, diethylolglycine, dihydroxyethylglycine, diethanol glycine, n,n-di 2-hydroxyethyl glycine, bicene, 2-bis 2-hydroxyethyl amino acetic acid, n,n-dihydroxyethyl glycine, n,n-dihydroxyethylglycine | |
| CHEBI:40957 | |
| [Na+].OCCN(CCO)CC([O-])=O |
Specifications
| 195°C | |
| White | |
| 25 g | |
| MFCD00004295 | |
| MFBDBXAVPLFMNJ-UHFFFAOYSA-M | |
| sodium 2-[bis(2-hydroxyethyl)amino]acetate | |
| 8761 | |
| 163.17 | |
| N,N-Di(2-hydroxyethyl)glycine [Good′s buffer component for biological research] |
| 150-25-4 | |
| Crystals or Powder | |
| C6H12NNaO4 | |
| bicine, n,n-bis 2-hydroxyethyl glycine, diethylolglycine, dihydroxyethylglycine, diethanol glycine, n,n-di 2-hydroxyethyl glycine, bicene, 2-bis 2-hydroxyethyl amino acetic acid, n,n-dihydroxyethyl glycine, n,n-dihydroxyethylglycine | |
| [Na+].OCCN(CCO)CC([O-])=O | |
| 185.16 | |
| CHEBI:40957 | |
| ≥99.0% (T) |
Safety and Handling
EINECSNumber : (2)-1287
RTECSNumber : MB9700000
TSCA : Yes