missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Di(2-hydroxyethyl)glycine 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 195°C |
| Color | White |
| Physical Form | Crystals or Powder |
| Quantity | 25 g |
| Formula Weight | 163.17 |
| Percent Purity | ≥99.0% (T) |
| Chemical Name or Material | N,N-Di(2-hydroxyethyl)glycine [Good′s buffer component for biological research] |
Safety and Handling
| EINECSNumber | (2)-1287 |
| RTECSNumber | MB9700000 |
| TSCA | Yes |