missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(acryloyl)cystamine, 98%

Catalog No. p-4408317
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4413203 1 g
AA4413206 5 g
AA4413214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4413203 Supplier Thermo Scientific Chemicals Supplier No. 04413203

Used as a reversible cross-linker for polyacrylamide gels | CAS: 60984-57-8 | C10H16N2O2S2 | 260.37 g/mol

N,N'-Bis(acryloyl)cystamine is used as a reversible cross-linker for polyacrylamide gels. It is also used for DNA purification. Further, it finds application in Suzuki reaction and fabrication of hydrogel modified plasmonic crystal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60984-57-8
Molecular Formula C10H16N2O2S2
Molecular Weight (g/mol) 260.37
MDL Number MFCD00036225
InChI Key DJVKJGIZQFBFGS-UHFFFAOYSA-N
Synonym n,n'-bis acryloyl cystamine, n,n'-bisacrylylcystamine, 2-propenamide, n,n'-dithiodi-2,1-ethanediyl bis, 2,2'-bisacrylamino diethyldisulfide, bis-acrylylcystamine: bac, n,n'-dithiodi-2,1-ethanediyl bis acrylamide, n-2-2-prop-2-enamido ethyl disulfanyl ethyl prop-2-enamide, n-2-2-prop-2-enoylamino ethyldisulfanyl ethyl prop-2-enamide, acmc-20ak9p, n,n-bis-acrylylcystamine
PubChem CID 100602
IUPAC Name N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide
SMILES C=CC(=O)NCCSSCCNC(=O)C=C

Specifications

Melting Point 123°C to 124°C
Assay Percent Range 98%
Quantity 1 g
Solubility Information Soluble in acetic acid. Slightly soluble in water.
Formula Weight 260.37
Physical Form Powder
Chemical Name or Material N, N′-Bis(acryloyl)cystamine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 262-546-8
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only