accessibility menu, dialog, popup

UserName

CAS RN 60984-57-8

IUPAC Name: N-(2-{[2-(prop-2-enamido)ethyl]disulfanyl}ethyl)prop-2-enamide
Synonyms: N-(2-{[2-(prop-2-enamido)ethyl]disulfanyl}ethyl)prop-2-enamide
Molecular Weight (g/mol): 260.37
Molecular Formula: C10H16N2O2S2
InChi Key: DJVKJGIZQFBFGS-UHFFFAOYSA-N
SMILES: C=CC(=O)NCCSSCCNC(=O)C=C
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results