missing translation for 'onlineSavingsMsg'
Learn More

N.N'-Bis(2-hydroxyethyl)oxamide, 99%

CAS: 1871-89-2 | C6H12N2O4 | 176.172 g/mol

Supplier:  Thermo Scientific Chemicals B2045814

 View more versions of this product

Catalog No. AAB2045814


Only null left
Explore available promotions

Description

Description

N.N'-Bis(2-hydroxyethyl)oxamide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N.N′-Bis(2-hydroxyethyl)oxamide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents.
Specifications

Specifications

N.N'-Bis(2-hydroxyethyl)oxamide
167°C to 169°C (decomposition)
MFCD00020559
Hygroscopic
Insoluble in water.
C(CO)NC(=O)C(=O)NCCO
176.172
176.17
1871-89-2
C6H12N2O4
25 g
n,n'-bis 2-hydroxyethyl oxamide, n1,n2-bis 2-hydroxyethyl oxalamide, nn'-bis 2-hydroxyethyl oxamide, unii-t92n2yr78x, ethanediamide, n,n'-bis 2-hydroxyethyl, n,n-bis 2-hydroxyethyl oxamide, n,n'-bis-2-hydroxyethyl-oxamide, ethanediamide, n1,n2-bis 2-hydroxyethyl, n,n'-bis 2-hydroxyethyl ethanediamide, n-2-hydroxyethyl-n'-2-hydroxyethyl ethane-1,2-diamide
FPQJEXTVQZHURJ-UHFFFAOYSA-N
N,N'-bis(2-hydroxyethyl)oxamide
74638
99%

Chemical Identifiers

1871-89-2
176.172
FPQJEXTVQZHURJ-UHFFFAOYSA-N
74638
C(CO)NC(=O)C(=O)NCCO
C6H12N2O4
MFCD00020559
n,n'-bis 2-hydroxyethyl oxamide, n1,n2-bis 2-hydroxyethyl oxalamide, nn'-bis 2-hydroxyethyl oxamide, unii-t92n2yr78x, ethanediamide, n,n'-bis 2-hydroxyethyl, n,n-bis 2-hydroxyethyl oxamide, n,n'-bis-2-hydroxyethyl-oxamide, ethanediamide, n1,n2-bis 2-hydroxyethyl, n,n'-bis 2-hydroxyethyl ethanediamide, n-2-hydroxyethyl-n'-2-hydroxyethyl ethane-1,2-diamide
N,N'-bis(2-hydroxyethyl)oxamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only