missing translation for 'onlineSavingsMsg'
Learn More

N.N'-Bis(2-hydroxyethyl)oxamide, 99%

Catalog No. p-7025121
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2045806 5 g
AAB2045814 25 g
AAB2045822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2045806 Supplier Thermo Scientific Chemicals Supplier No. B2045806

CAS: 1871-89-2 | C6H12N2O4 | 176.172 g/mol

N.N'-Bis(2-hydroxyethyl)oxamide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N.N′-Bis(2-hydroxyethyl)oxamide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 1871-89-2
Molecular Formula C6H12N2O4
Molecular Weight (g/mol) 176.172
MDL Number MFCD00020559
InChI Key FPQJEXTVQZHURJ-UHFFFAOYSA-N
Synonym n,n'-bis 2-hydroxyethyl oxamide, n1,n2-bis 2-hydroxyethyl oxalamide, nn'-bis 2-hydroxyethyl oxamide, unii-t92n2yr78x, ethanediamide, n,n'-bis 2-hydroxyethyl, n,n-bis 2-hydroxyethyl oxamide, n,n'-bis-2-hydroxyethyl-oxamide, ethanediamide, n1,n2-bis 2-hydroxyethyl, n,n'-bis 2-hydroxyethyl ethanediamide, n-2-hydroxyethyl-n'-2-hydroxyethyl ethane-1,2-diamide
PubChem CID 74638
IUPAC Name N,N'-bis(2-hydroxyethyl)oxamide
SMILES C(CO)NC(=O)C(=O)NCCO

Specifications

Melting Point 167°C to 169°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 176.17
Percent Purity 99%
Chemical Name or Material N.N'-Bis(2-hydroxyethyl)oxamide
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only