accessibility menu, dialog, popup

UserName

HOHNOONHOH

CAS RN 1871-89-2

IUPAC Name: N,N'-bis(2-hydroxyethyl)ethanediamide
Synonyms: N,N'-bis(2-hydroxyethyl)ethanediamide
Molecular Weight (g/mol): 176.17
Molecular Formula: C6H12N2O4
InChi Key: FPQJEXTVQZHURJ-UHFFFAOYSA-N
SMILES: OCCNC(=O)C(=O)NCCO
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)