missing translation for 'onlineSavingsMsg'
Learn More

N-Methylbenzylamine 98.0+%, TCI America™

Catalog No. M016425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M016425ML 25 mL
M0164500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M016425ML Supplier TCI America Supplier No. M016425ML

Chemical Identifiers

CAS 103-67-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008289
InChI Key RIWRFSMVIUAEBX-UHFFFAOYSA-N
Synonym n-methylbenzylamine, methylbenzylamine, n-benzylmethylamine, benzyl methyl amine, benzenemethanamine, n-methyl, benzylmethylamine, n-methy-n-benzylamine, n-benzyl-n-methylamine, benzylamine, n-methyl, n-methyl phenyl methanamine
PubChem CID 7669
IUPAC Name benzyl(methyl)amine
SMILES CNCC1=CC=CC=C1

Specifications

Melting Point -55°C
Boiling Point 186°C
Quantity 25 mL
UN Number 2735
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methylbenzylamine
EINECSNumber (3)-4480
TSCA Yes