accessibility menu, dialog, popup

UserName

CH3HN

CAS RN 103-67-3

IUPAC Name: benzyl(methyl)amine
Synonyms: N-methylbenzylamine
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: RIWRFSMVIUAEBX-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)