missing translation for 'onlineSavingsMsg'
Learn More

N-Methylbenzylamine, 97%

Catalog No. p-160293
Change view
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106040050 5 mL Glass bottle
AC106045000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106040050 Supplier Thermo Scientific Chemicals Supplier No. 106040050

CAS: 103-67-3 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-67-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008289
InChI Key RIWRFSMVIUAEBX-UHFFFAOYSA-N
Synonym n-methylbenzylamine, methylbenzylamine, n-benzylmethylamine, benzyl methyl amine, benzenemethanamine, n-methyl, benzylmethylamine, n-methy-n-benzylamine, n-benzyl-n-methylamine, benzylamine, n-methyl, n-methyl phenyl methanamine
PubChem CID 7669
IUPAC Name N-methyl-1-phenylmethanamine
SMILES CNCC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -24°C
Color Colorless to Yellow
Density 0.9400g/mL
Boiling Point 184°C to 189°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5214 to 1.5234
Linear Formula C6H5CH2NHCH3
Quantity 5 mL
Beilstein 12, 1019
Specific Gravity 0.94
Solubility Information Solubility in water: 65g/L (20°C)
Formula Weight 121.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Methylbenzylamine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 203-133-4
TSCA TSCA

RUO – Research Use Only