missing translation for 'onlineSavingsMsg'
Learn More

N-Methylaniline, 98%

CAS: 100-61-8 | C7H9N | 107.16 g/mol

Supplier:  Thermo Scientific Chemicals L138730E

 View more versions of this product

Catalog No. AAL138730E

Only null left
Explore available promotions

Description

Description

N-Methylaniline is used in the preparation of N,N'-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylaniline is used in the preparation of N,N′-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides, acid chlorides and carbon dioxide.
Specifications

Specifications

N-Methylaniline
-57°C
194°C to 197°C
C7H9N
MFCD00008283
UN2294
Air Sensitive
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
AFBPFSWMIHJQDM-UHFFFAOYSA-N
N-methylaniline
7515
107.16
100-61-8
0.989
86°C (186°F)
1.571
2500 g
741982
14,6019
Immiscible with water.
CNC1=CC=CC=C1
107.16
CHEBI:15733
98%

Chemical Identifiers

100-61-8
107.16
AFBPFSWMIHJQDM-UHFFFAOYSA-N
7515
N-methylaniline
C7H9N
MFCD00008283
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
CHEBI:15733
CNC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c

H227-H301+H311+H331-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N-METHYLANILINE

EINECSNumber : 202-870-9

RTECSNumber : BY4550000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only