accessibility menu, dialog, popup

UserName

CAS RN 100-61-8

IUPAC Name: N-methylaniline
Synonyms: methylaniline
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: AFBPFSWMIHJQDM-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)