Learn More
N-Methylaniline, 98%
Description
N-Methylaniline is used in the preparation of N,N'-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Methylaniline is used in the preparation of N,N′-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides, acid chlorides and carbon dioxide.
Specifications
Specifications
| Melting Point | -57°C |
| Density | 0.989 |
| Boiling Point | 194°C to 197°C |
| Flash Point | 86°C (186°F) |
| Refractive Index | 1.571 |
| Quantity | 100 g |
| UN Number | UN2294 |
| Beilstein | 741982 |
| Sensitivity | Air Sensitive |
| Merck Index | 14,6019 |
| Show More |
Safety and Handling
| Hazard Category | H227-H301+H311+H331-H373 |
| Hazard Statement | GHS H Statement H301-H311-H331-H373-H227 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Combustible liquid. |
| Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N-METHYLANILINE |
| EINECSNumber | 202-870-9 |
| RTECSNumber | BY4550000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only