missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-p-anisidine 96.0+%, TCI America™

Catalog No. M17431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17431G 1 g
M17435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17431G Supplier TCI America Supplier No. M17431G

Chemical Identifiers

CAS 5961-59-1
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00008399
InChI Key JFXDIXYFXDOZIT-UHFFFAOYSA-N
Synonym n-methyl-p-anisidine, n-methyl-4-anisidine, n-methyl-4-methoxyaniline, benzenamine, 4-methoxy-n-methyl, n-methyl-p-ansidine, p-anisidine, n-methyl, 4-methoxy-n-methylbenzenamine, 1-methylamino-4-methoxybenzene, p-methoxy-n-methylaniline, acmc-209mes
PubChem CID 22250
IUPAC Name 4-methoxy-N-methylaniline
SMILES CNC1=CC=C(C=C1)OC

Specifications

Melting Point 35°C
Boiling Point 136°C
Quantity 1 g
Formula Weight 137.18
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-p-anisidine