missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-p-anisidine, 98%

Catalog No. p-160217
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC177220010 1 g Glass bottle
AC177220100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC177220010 Supplier Thermo Scientific Chemicals Supplier No. 177220010

CAS: 5961-59-1 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5961-59-1
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008399
InChI Key JFXDIXYFXDOZIT-UHFFFAOYSA-N
Synonym n-methyl-p-anisidine, n-methyl-4-anisidine, n-methyl-4-methoxyaniline, benzenamine, 4-methoxy-n-methyl, n-methyl-p-ansidine, p-anisidine, n-methyl, 4-methoxy-n-methylbenzenamine, 1-methylamino-4-methoxybenzene, p-methoxy-n-methylaniline, acmc-209mes
PubChem CID 22250
IUPAC Name 4-methoxy-N-methylaniline
SMILES CNC1=CC=C(C=C1)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33.0°C to 36.0°C
Color Brown-Gray to White
Boiling Point 135.0°C to 136.0°C (19.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC6H4NHCH3
Quantity 1 g
Beilstein 13, 442
Formula Weight 137.18
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material N-Methyl-p-anisidine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/va
EINECSNumber 227-735-1

RUO – Research Use Only