missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-propargylbenzylamine, 97%

CAS: 555-57-7 | C11H13N | 159.23 g/mol

$66.79 - $152.37

Chemical Identifiers

CAS 555-57-7
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.23
MDL Number MFCD00008576
InChI Key DPWPWRLQFGFJFI-UHFFFAOYSA-N
Synonym pargyline, pargylamine, n-methyl-n-propargylbenzylamine, paragyline, eudatin, supirdyl, eutonyl, pargylin, n-methyl-n-2-propynylbenzylamine, benzyl-methyl-2-propinylamin
PubChem CID 4688
ChEBI CHEBI:7930
IUPAC Name N-benzyl-N-methylprop-2-yn-1-amine
SMILES CN(CC#C)CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC127530010
SDS View Documents
Thermo Scientific Chemicals
127530010
1 g Glass bottle
Each for $66.79
Only null left
 
AC127530050
SDS View Documents
Thermo Scientific Chemicals
127530050
5 g Glass bottle
Each for $152.37
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

555-57-7
159.23
DPWPWRLQFGFJFI-UHFFFAOYSA-N
4688
N-benzyl-N-methylprop-2-yn-1-amine
C11H13N
MFCD00008576
pargyline, pargylamine, n-methyl-n-propargylbenzylamine, paragyline, eudatin, supirdyl, eutonyl, pargylin, n-methyl-n-2-propynylbenzylamine, benzyl-methyl-2-propinylamin
CHEBI:7930
CN(CC#C)CC1=CC=CC=C1

Specifications

555-57-7
100.0
0.9400g/mL
83°C
97%
C11H13N
C6H5CH2N(CH3)CH2C∽CH
1 g
0.94
pargyline, pargylamine, n-methyl-n-propargylbenzylamine, paragyline, eudatin, supirdyl, eutonyl, pargylin, n-methyl-n-2-propynylbenzylamine, benzyl-methyl-2-propinylamin
DPWPWRLQFGFJFI-UHFFFAOYSA-N
N-benzyl-N-methylprop-2-yn-1-amine
4688
159.23
Liquid
96.0
Yellow
86°C to 88°C (4.0 mmHg)
Authentic
Glass bottle
1.5213
MFCD00008576
12, II, 548
15, 7142
Solubility in water: soluble. Other solubilities: miscibel in ethanol,acetone
CN(CC#C)CC1=CC=CC=C1
159.23
CHEBI:7930
97%
N-Methyl-N-propargylbenzylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-101-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only