accessibility menu, dialog, popup

UserName

CAS RN 555-57-7

IUPAC Name: benzyl(methyl)(prop-2-yn-1-yl)amine
Synonyms: pargyline pargyline preparation pargyline (substance) pargylin paragyline pargyline (product) pargylinum pargilina benzyl-methyl-2-propinylamin methylbenzylpropynylamine
Molecular Weight (g/mol): 159.23
Molecular Formula: C11H13N
InChi Key: DPWPWRLQFGFJFI-UHFFFAOYSA-N
SMILES: CN(CC#C)CC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)