missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-m-toluidine 98.0+%, TCI America™
Supplier: TCI America M0446100ML
Specifications
| N-Methyl-m-toluidine | |
| Yellow | |
| C8H11N | |
| 100 mL | |
| n-methyl-m-toluidine, benzenamine, n,3-dimethyl, 3-methylamino toluene, m,n-dimethylaniline, m-toluidine, n-methyl, n-methyl-3-methylaniline, n-3-dimethylbenzenamine, aniline, n,m-dimethyl, 3,n-dimethylaniline, n-methyl-3-toluidine | |
| CC1=CC(=CC=C1)NC | |
| 121.183 | |
| 121.18 | |
| Liquid |
| 696-44-6 | |
| 207°C | |
| MFCD00035787 | |
| 2810 | |
| FBGJJTQNZVNEQU-UHFFFAOYSA-N | |
| N,3-dimethylaniline | |
| 69675 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 696-44-6 | |
| 121.183 | |
| FBGJJTQNZVNEQU-UHFFFAOYSA-N | |
| 69675 | |
| CC1=CC(=CC=C1)NC |
| C8H11N | |
| MFCD00035787 | |
| n-methyl-m-toluidine, benzenamine, n,3-dimethyl, 3-methylamino toluene, m,n-dimethylaniline, m-toluidine, n-methyl, n-methyl-3-methylaniline, n-3-dimethylbenzenamine, aniline, n,m-dimethyl, 3,n-dimethylaniline, n-methyl-3-toluidine | |
| N,3-dimethylaniline |
Safety and Handling
EINECSNumber : (3)-0129&(3)-0187
TSCA : Yes