missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-m-toluidine 98.0+%, TCI America™

Catalog No. p-7132177
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M044625ML 25 mL
M0446100ML 100 mL
M0446500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M044625ML Supplier TCI America Supplier No. M044625ML
Only null left

Chemical Identifiers

CAS 696-44-6
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00035787
InChI Key FBGJJTQNZVNEQU-UHFFFAOYSA-N
Synonym n-methyl-m-toluidine, benzenamine, n,3-dimethyl, 3-methylamino toluene, m,n-dimethylaniline, m-toluidine, n-methyl, n-methyl-3-methylaniline, n-3-dimethylbenzenamine, aniline, n,m-dimethyl, 3,n-dimethylaniline, n-methyl-3-toluidine
PubChem CID 69675
IUPAC Name N,3-dimethylaniline
SMILES CC1=CC(=CC=C1)NC

Specifications

Color Yellow
Boiling Point 207°C
Quantity 25 mL
UN Number 2810
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methyl-m-toluidine
EINECSNumber (3)-0129&(3)-0187
TSCA Yes