missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-D-glucamine Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America M071325G

 View more versions of this product

Catalog No. M071325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Methyl-D-glucamine Hydrochloride [for Buffer]
150°C
C7H18ClNO5
25 g
CNCC(C(C(C(CO)O)O)O)O.Cl
231.673
231.67
Crystalline Powder
35564-86-4
White
MFCD00060164
PKPZZAVJXDZHDW-LJTMIZJLSA-N
(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;hydrochloride
44630381
≥98.0% (HPLC,T)

Chemical Identifiers

35564-86-4
231.673
PKPZZAVJXDZHDW-LJTMIZJLSA-N
(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;hydrochloride
C7H18ClNO5
MFCD00060164
44630381
CNCC(C(C(C(CO)O)O)O)O.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions