accessibility menu, dialog, popup

UserName

CAS RN 35564-86-4

H3CNHOHOHOHOHOHHCl

CAS RN 35564-86-4

IUPAC Name: 6-(methylamino)hexane-1,2,3,4,5-pentol hydrochloride
Synonyms: methylglucamine hcl
Molecular Weight (g/mol): 231.67
Molecular Formula: C7H18ClNO5
InChi Key: PKPZZAVJXDZHDW-UHFFFAOYNA-N
SMILES: Cl.CNCC(O)C(O)C(O)C(O)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)