missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-D-glucamine Hydrochloride 98.0+%, TCI America™

Catalog No. M07135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07135G 5 g
M071325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07135G Supplier TCI America Supplier No. M07135G
Only null left

Chemical Identifiers

CAS 35564-86-4
Molecular Formula C7H18ClNO5
Molecular Weight (g/mol) 231.673
MDL Number MFCD00060164
InChI Key PKPZZAVJXDZHDW-LJTMIZJLSA-N
PubChem CID 44630381
IUPAC Name (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;hydrochloride
SMILES CNCC(C(C(C(CO)O)O)O)O.Cl

Specifications

Melting Point 150°C
Color White
Quantity 5 g
Formula Weight 231.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-D-glucamine Hydrochloride [for Buffer]