missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Methyl-D-glucamine, 99%

CAS: 6284-40-8 | C7H17NO5 | 195.215 g/mol

Supplier:  Thermo Scientific Chemicals L1428230

 View more versions of this product

Catalog No. AAL1428230


Only null left
Explore available promotions

Description

Description

N-Methyl-D-glucamine is used in conjunction with iodinated organic compounds in contrast media viz. diatrizoate meglumine and iodipamide meglumine. It is useful in ion-exchange resins to chelate boron ions selectively. It is also used as an excipient in pharmaceuticals, in cosmetics and X-ray contrast media. It is also employed in the synthesis of surface active agents, pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyl-D-glucamine is used in conjunction with iodinated organic compounds in contrast media viz. diatrizoate meglumine and iodipamide meglumine. It is useful in ion-exchange resins to chelate boron ions selectively. It is also used as an excipient in pharmaceuticals, in cosmetics and X-ray contrast media. It is also employed in the synthesis of surface active agents, pharmaceuticals and dyes.

Solubility
Soluble in water.

Notes
Air sensitive and hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

N-Methyl-D-glucamine
128°C to 131°C
MFCD00004707
385906
14,6078
Soluble in water.
−16.5° (c=2 in Water)
(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol
8567
195.22
6284-40-8
C7H17NO5
250 g
Air sensitive; Hygroscopic
meglumine, n-methyl-d-glucamine, n-methylglucamine, meglumin, 1-deoxy-1-methylamino-d-glucitol, methylglucamin, meglumina, 1-deoxy-1-methylaminosorbitol, d-glucitol, 1-deoxy-1-methylamino, n-methyl-d--glucamine
MBBZMMPHUWSWHV-BDVNFPICSA-N
CNCC(C(C(C(CO)O)O)O)O
195.215
CHEBI:59732
99%

Chemical Identifiers

6284-40-8
195.215
MBBZMMPHUWSWHV-BDVNFPICSA-N
8567
(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol
C7H17NO5
MFCD00004707
meglumine, n-methyl-d-glucamine, n-methylglucamine, meglumin, 1-deoxy-1-methylamino-d-glucitol, methylglucamin, meglumina, 1-deoxy-1-methylaminosorbitol, d-glucitol, 1-deoxy-1-methylamino, n-methyl-d--glucamine
CHEBI:59732
CNCC(C(C(C(CO)O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 228-506-9

RTECSNumber : LZ4295770

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only