missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Methyl-D-glucamine, 99%

Catalog No. p-7046421
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1428218 50 g
AAL1428230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1428218 Supplier Thermo Scientific Chemicals Supplier No. L1428218
Only null left

CAS: 6284-40-8 | C7H17NO5 | 195.215 g/mol

N-Methyl-D-glucamine is used in conjunction with iodinated organic compounds in contrast media viz. diatrizoate meglumine and iodipamide meglumine. It is useful in ion-exchange resins to chelate boron ions selectively. It is also used as an excipient in pharmaceuticals, in cosmetics and X-ray contrast media. It is also employed in the synthesis of surface active agents, pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyl-D-glucamine is used in conjunction with iodinated organic compounds in contrast media viz. diatrizoate meglumine and iodipamide meglumine. It is useful in ion-exchange resins to chelate boron ions selectively. It is also used as an excipient in pharmaceuticals, in cosmetics and X-ray contrast media. It is also employed in the synthesis of surface active agents, pharmaceuticals and dyes.

Solubility
Soluble in water.

Notes
Air sensitive and hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 6284-40-8
Molecular Formula C7H17NO5
Molecular Weight (g/mol) 195.215
MDL Number MFCD00004707
InChI Key MBBZMMPHUWSWHV-BDVNFPICSA-N
Synonym meglumine, n-methyl-d-glucamine, n-methylglucamine, meglumin, 1-deoxy-1-methylamino-d-glucitol, methylglucamin, meglumina, 1-deoxy-1-methylaminosorbitol, d-glucitol, 1-deoxy-1-methylamino, n-methyl-d--glucamine
PubChem CID 8567
ChEBI CHEBI:59732
IUPAC Name (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol
SMILES CNCC(C(C(C(CO)O)O)O)O

Specifications

Melting Point 128°C to 131°C
Quantity 50 g
Beilstein 385906
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,6078
Solubility Information Soluble in water.
Optical Rotation −16.5° (c=2 in Water)
Formula Weight 195.22
Percent Purity 99%
Chemical Name or Material N-Methyl-D-glucamine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 228-506-9
RTECSNumber LZ4295770
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only