missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(5-phenylpyrid-2-yl)methylamine 97%, Thermo Scientific™

Chemical Identifiers

CAS 892502-02-2
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
MDL Number MFCD10700051
InChI Key HHOWXRCPAWWJPH-UHFFFAOYSA-N
Synonym n-methyl 5-phenylpyrid-2-yl methylamine, methyl 5-phenylpyridin-2-yl methyl amine, 2-methyl aminomethyl-5-phenylpyridine, 2-pyridinemethanamine,n-methyl-5-phenyl, methyl 5-phenyl 2-pyridyl methyl amine, n-methyl-5-phenylpyridin-2-yl methylamine, n-methyl-1-5-phenylpyridin-2-yl methanamine
PubChem CID 26343606
IUPAC Name N-methyl-1-(5-phenylpyridin-2-yl)methanamine
SMILES CNCC1=NC=C(C=C1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC55046CB
SDS View Documents
Thermo Scientific Chemicals
CC55046CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC55046DA
SDS View Documents
Thermo Scientific Chemicals
CC55046DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

892502-02-2
198.269
HHOWXRCPAWWJPH-UHFFFAOYSA-N
26343606
CNCC1=NC=C(C=C1)C2=CC=CC=C2
C13H14N2
MFCD10700051
n-methyl 5-phenylpyrid-2-yl methylamine, methyl 5-phenylpyridin-2-yl methyl amine, 2-methyl aminomethyl-5-phenylpyridine, 2-pyridinemethanamine,n-methyl-5-phenyl, methyl 5-phenyl 2-pyridyl methyl amine, n-methyl-5-phenylpyridin-2-yl methylamine, n-methyl-1-5-phenylpyridin-2-yl methanamine
N-methyl-1-(5-phenylpyridin-2-yl)methanamine

Specifications

892502-02-2
100.0
Amber Glass Bottle
MFCD10700051
n-methyl 5-phenylpyrid-2-yl methylamine, methyl 5-phenylpyridin-2-yl methyl amine, 2-methyl aminomethyl-5-phenylpyridine, 2-pyridinemethanamine,n-methyl-5-phenyl, methyl 5-phenyl 2-pyridyl methyl amine, n-methyl-5-phenylpyridin-2-yl methylamine, n-methyl-1-5-phenylpyridin-2-yl methanamine
CNCC1=NC=C(C=C1)C2=CC=CC=C2
198.269
198.27
n-methyl(5-phenylpyrid-2-yl)methylamine
89.5
97%
C13H14N2
250mg
HHOWXRCPAWWJPH-UHFFFAOYSA-N
N-methyl-1-(5-phenylpyridin-2-yl)methanamine
26343606
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents