missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(5-phenylpyrid-2-yl)methylamine 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC55046DA

 View more versions of this product

Catalog No. CC55046DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

n-methyl(5-phenylpyrid-2-yl)methylamine
89.5
97%
C13H14N2
1g
HHOWXRCPAWWJPH-UHFFFAOYSA-N
N-methyl-1-(5-phenylpyridin-2-yl)methanamine
26343606
90%
892502-02-2
100.0
Amber Glass Bottle
MFCD10700051
n-methyl 5-phenylpyrid-2-yl methylamine, methyl 5-phenylpyridin-2-yl methyl amine, 2-methyl aminomethyl-5-phenylpyridine, 2-pyridinemethanamine,n-methyl-5-phenyl, methyl 5-phenyl 2-pyridyl methyl amine, n-methyl-5-phenylpyridin-2-yl methylamine, n-methyl-1-5-phenylpyridin-2-yl methanamine
CNCC1=NC=C(C=C1)C2=CC=CC=C2
198.269
198.27

Chemical Identifiers

892502-02-2
198.269
HHOWXRCPAWWJPH-UHFFFAOYSA-N
26343606
CNCC1=NC=C(C=C1)C2=CC=CC=C2
C13H14N2
MFCD10700051
n-methyl 5-phenylpyrid-2-yl methylamine, methyl 5-phenylpyridin-2-yl methyl amine, 2-methyl aminomethyl-5-phenylpyridine, 2-pyridinemethanamine,n-methyl-5-phenyl, methyl 5-phenyl 2-pyridyl methyl amine, n-methyl-5-phenylpyridin-2-yl methylamine, n-methyl-1-5-phenylpyridin-2-yl methanamine
N-methyl-1-(5-phenylpyridin-2-yl)methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions