CAS RN 892502-02-2
CAS RN 892502-02-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Methyl(5-phenylpyrid-2-yl)methylamine 97%, Thermo Scientific™
CAS: 892502-02-2 Molecular Formula: C13H14N2 Molecular Weight (g/mol): 198.269 MDL Number: MFCD10700051 InChI Key: HHOWXRCPAWWJPH-UHFFFAOYSA-N Synonym: n-methyl 5-phenylpyrid-2-yl methylamine,methyl 5-phenylpyridin-2-yl methyl amine,2-methyl aminomethyl-5-phenylpyridine,2-pyridinemethanamine,n-methyl-5-phenyl,methyl 5-phenyl 2-pyridyl methyl amine,n-methyl-5-phenylpyridin-2-yl methylamine,n-methyl-1-5-phenylpyridin-2-yl methanamine PubChem CID: 26343606 IUPAC Name: N-methyl-1-(5-phenylpyridin-2-yl)methanamine SMILES: CNCC1=NC=C(C=C1)C2=CC=CC=C2