missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-4-(trifluoromethyl)aniline 98.0+%, TCI America™
$190.13 - $480.92
Chemical Identifiers
| CAS | 22864-65-9 |
|---|---|
| Molecular Formula | C8H8F3N |
| Molecular Weight (g/mol) | 175.154 |
| MDL Number | MFCD07369717 |
| InChI Key | UTUYWZJPVLDHJJ-UHFFFAOYSA-N |
| PubChem CID | 15099755 |
| IUPAC Name | N-methyl-4-(trifluoromethyl)aniline |
| SMILES | CNC1=CC=C(C=C1)C(F)(F)F |
Chemical Identifiers
| 22864-65-9 | |
| 175.154 | |
| UTUYWZJPVLDHJJ-UHFFFAOYSA-N | |
| N-methyl-4-(trifluoromethyl)aniline |
| C8H8F3N | |
| MFCD07369717 | |
| 15099755 | |
| CNC1=CC=C(C=C1)C(F)(F)F |
Specifications
| 22864-65-9 | |
| 106°C | |
| MFCD07369717 | |
| UTUYWZJPVLDHJJ-UHFFFAOYSA-N | |
| N-methyl-4-(trifluoromethyl)aniline | |
| 15099755 | |
| ≥98.0% (GC,T) | |
| N-Methyl-4-(trifluoromethyl)aniline |
| Yellow | |
| C8H8F3N | |
| 1 g | |
| CNC1=CC=C(C=C1)C(F)(F)F | |
| 175.154 | |
| 175.15 | |
| Liquid |
Safety and Handling
TSCA : No