CAS RN 22864-65-9
IUPAC Name:
N-methyl-4-(trifluoromethyl)aniline
Synonyms:
N-methyl-4-(trifluoromethyl)aniline
Molecular Weight (g/mol):
175.15
Molecular Formula:
C8H8F3N
InChi Key:
UTUYWZJPVLDHJJ-UHFFFAOYSA-N
SMILES:
CNC1=CC=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
N-Methyl-4-(trifluoromethyl)aniline 98.0+%, TCI America™
CAS: 22864-65-9 Molecular Formula: C8H8F3N Molecular Weight (g/mol): 175.154 MDL Number: MFCD07369717 InChI Key: UTUYWZJPVLDHJJ-UHFFFAOYSA-N PubChem CID: 15099755 IUPAC Name: N-methyl-4-(trifluoromethyl)aniline SMILES: CNC1=CC=C(C=C1)C(F)(F)F