missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-4-(trifluoromethyl)aniline 98.0+%, TCI America™

Catalog No. M27741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27741G 1 g
M27745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27741G Supplier TCI America Supplier No. M27741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22864-65-9
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD07369717
InChI Key UTUYWZJPVLDHJJ-UHFFFAOYSA-N
PubChem CID 15099755
IUPAC Name N-methyl-4-(trifluoromethyl)aniline
SMILES CNC1=CC=C(C=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 106°C
Quantity 1 g
Formula Weight 175.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methyl-4-(trifluoromethyl)aniline