missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-4-nitro-N-[2-(4-nitrophenoxy)ethyl]phenylethylamine 97.0+%, TCI America™

Catalog No. M19991G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19991G Supplier TCI America Supplier No. M19991G

Chemical Identifiers

CAS 115287-37-1
Molecular Formula C17H19N3O5
Molecular Weight (g/mol) 345.36
MDL Number MFCD08236712
InChI Key BDCVRCLAXLOSLH-UHFFFAOYSA-N
PubChem CID 14645090
IUPAC Name methyl[2-(4-nitrophenoxy)ethyl][2-(4-nitrophenyl)ethyl]amine
SMILES CN(CCOC1=CC=C(C=C1)[N+]([O-])=O)CCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 72°C
Quantity 1 g
Formula Weight 345.36
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-4-nitro-N-[2-(4-nitrophenoxy)ethyl]phenylethylamine