accessibility menu, dialog, popup

UserName

CAS RN 115287-37-1

CH3NNOOONOO

CAS RN 115287-37-1

IUPAC Name: methyl[2-(4-nitrophenoxy)ethyl][2-(4-nitrophenyl)ethyl]amine
Synonyms: methyl[2-(4-nitrophenoxy)ethyl][2-(4-nitrophenyl)ethyl]amine
Molecular Weight (g/mol): 345.36
Molecular Formula: C17H19N3O5
InChi Key: BDCVRCLAXLOSLH-UHFFFAOYSA-N
SMILES: CN(CCOC1=CC=C(C=C1)[N+]([O-])=O)CCC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results