missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-4-methylbenzamide, 97%, Thermo Scientific™

Catalog No. p-7037659
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56446MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH56446MD Supplier Thermo Scientific Chemicals Supplier No. H56446MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 18370-11-1
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00043627
InChI Key FZIOOTTWDRFBKU-UHFFFAOYSA-N
Synonym n-methyl-p-toluamide, n,p-dimethylbenzamide, n-methyl 4-methylphenyl carboxamide, n-methyl-4-toluamide, n,4-dimethyl-benzamide, n-methyl-p-methylbenzamide, 4-methyl-n-methylbenzamide, n-methyl 4-methylbenzamide, n-methyl-4-methylbenzamide
PubChem CID 140377
IUPAC Name N,4-dimethylbenzamide
SMILES CC1=CC=C(C=C1)C(=O)NC

Specifications

Quantity 250 mg
Formula Weight 149.18
Percent Purity 97%
Chemical Name or Material N-Methyl-4-methylbenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures