missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-(1-methyl-1h-indol-7-yl)methylamine, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC66946DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC66946DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

709649-75-2
174.247
JERMRTDADLXHSD-UHFFFAOYSA-N
33589520
CNCC1=CC=CC2=C1N(C=C2)C
C11H14N2
MFCD11506353
methyl 1-methylindol-7-yl methyl amine, n-methyl-1-1-methyl-1h-indol-7-yl methylamine, n-methyl-1-methyl-1h-indol-7-yl methylamine, 1h-indole-7-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-7-yl methanamine, 1h-indole-7-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-7-ylmethyl amine, 7-methylamino methyl-1-methyl-1h-indole, methyl 1-methyl-1h-indol-7-yl methyl amine
N-methyl-1-(1-methylindol-7-yl)methanamine

Specifications

N-methyl-(1-methyl-1H-indol-7-yl)methylamine
89.5
90%
C11H14N2
MFCD11506353
JERMRTDADLXHSD-UHFFFAOYSA-N
N-methyl-1-(1-methylindol-7-yl)methanamine
33589520
90%
709649-75-2
100.0
Amber Glass Bottle
1g
methyl 1-methylindol-7-yl methyl amine, n-methyl-1-1-methyl-1h-indol-7-yl methylamine, n-methyl-1-methyl-1h-indol-7-yl methylamine, 1h-indole-7-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-7-yl methanamine, 1h-indole-7-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-7-ylmethyl amine, 7-methylamino methyl-1-methyl-1h-indole, methyl 1-methyl-1h-indol-7-yl methyl amine
CNCC1=CC=CC2=C1N(C=C2)C
174.247
174.25
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents