missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-(1-methyl-1h-indol-7-yl)methylamine, 90%, Thermo Scientific™

Catalog No. p-3881617
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC66946CB 250mg Amber Glass Bottle
CC66946DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC66946CB Supplier Thermo Scientific Chemicals Supplier No. CC66946CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 709649-75-2
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.247
MDL Number MFCD11506353
InChI Key JERMRTDADLXHSD-UHFFFAOYSA-N
Synonym methyl 1-methylindol-7-yl methyl amine, n-methyl-1-1-methyl-1h-indol-7-yl methylamine, n-methyl-1-methyl-1h-indol-7-yl methylamine, 1h-indole-7-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-7-yl methanamine, 1h-indole-7-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-7-ylmethyl amine, 7-methylamino methyl-1-methyl-1h-indole, methyl 1-methyl-1h-indol-7-yl methyl amine
PubChem CID 33589520
IUPAC Name N-methyl-1-(1-methylindol-7-yl)methanamine
SMILES CNCC1=CC=CC2=C1N(C=C2)C

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 174.25
Percent Purity 90%
Chemical Name or Material N-methyl-(1-methyl-1H-indol-7-yl)methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.