accessibility menu, dialog, popup

UserName

CAS RN 709649-75-2

IUPAC Name: methyl[(1-methyl-1H-indol-7-yl)methyl]amine
Synonyms: methyl[(1-methylindol-7-yl)methyl]amine
Molecular Weight (g/mol): 174.25
Molecular Formula: C11H14N2
InChi Key: JERMRTDADLXHSD-UHFFFAOYSA-N
SMILES: CNCC1=C2N(C)C=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)