missing translation for 'onlineSavingsMsg'
Learn More

N-(m-Toluoyl)glycine 98.0+%, TCI America™

Supplier:  TCI America T031225G

 View more versions of this product

Catalog No. T031225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-(m-Toluoyl)glycine
138°C
C10H11NO3
25 g
YKAKNMHEIJUKEX-UHFFFAOYSA-N
2-[(3-methylphenyl)formamido]acetic acid
99223
193.20
Crystalline Powder
27115-49-7
White
MFCD00044399
3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid
CC1=CC=CC(=C1)C(=O)NCC(O)=O
193.20
CHEBI:68500
≥98.0% (T)

Chemical Identifiers

27115-49-7
193.20
YKAKNMHEIJUKEX-UHFFFAOYSA-N
99223
2-[(3-methylphenyl)formamido]acetic acid
C10H11NO3
MFCD00044399
3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid
CHEBI:68500
CC1=CC=CC(=C1)C(=O)NCC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions