missing translation for 'onlineSavingsMsg'
Learn More

N-(m-Toluoyl)glycine 98.0+%, TCI America™

$119.30 - $339.25

Chemical Identifiers

CAS 27115-49-7
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00044399
InChI Key YKAKNMHEIJUKEX-UHFFFAOYSA-N
Synonym 3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid
PubChem CID 99223
ChEBI CHEBI:68500
IUPAC Name 2-[(3-methylphenyl)formamido]acetic acid
SMILES CC1=CC=CC(=C1)C(=O)NCC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T03125G
View Documents
TCI America
T03125G
5 g
Each for $119.30
Only null left
 
T031225G
View Documents
TCI America
T031225G
25 g
Each for $339.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

27115-49-7
193.20
YKAKNMHEIJUKEX-UHFFFAOYSA-N
99223
2-[(3-methylphenyl)formamido]acetic acid
C10H11NO3
MFCD00044399
3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid
CHEBI:68500
CC1=CC=CC(=C1)C(=O)NCC(O)=O

Specifications

27115-49-7
White
MFCD00044399
3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid
CC1=CC=CC(=C1)C(=O)NCC(O)=O
193.20
CHEBI:68500
≥98.0% (T)
N-(m-Toluoyl)glycine
138°C
C10H11NO3
5 g
YKAKNMHEIJUKEX-UHFFFAOYSA-N
2-[(3-methylphenyl)formamido]acetic acid
99223
193.20
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'