missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine Sodium Salt, 94%, Spectrum™ Chemical

$183.07 - $5,995.64

Chemical Identifiers

CAS 137-16-6
Molecular Weight (g/mol) 293.39
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18612030
View Documents Promotion Details
N/A 25 g Poly Bottle
Each for $183.07
Only null left
 
18612031
View Documents Promotion Details
N/A 100 g Poly Bottle
Each for $378.79
Only null left
 
18612032
View Documents Promotion Details
N/A 500 g Poly Bottle
Each for $874.25
Only null left
 
18612033
View Documents Promotion Details
N/A 2.5 kg Poly Bottle
Each for $2,565.04
Only null left
 
18612034
View Documents Promotion Details
N/A 12 kg Poly Bottle
Each for $5,995.64
Only null left
 
Description

Description

Specifications

Chemical Identifiers

137-16-6
293.39

Specifications

137-16-6
7.5 to 8.5
94%
N to Lauroylsarcosine Sodium Salt
1
293.39
Ungraded
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.