missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine Sodium Salt, 94%, Spectrum™ Chemical

Catalog No. 18612034
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612034 12 kg Poly Bottle
18612030 25 g Poly Bottle
18612031 100 g Poly Bottle
18612032 500 g Poly Bottle
18612033 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612034 Supplier Spectrum Chemical Mfg Cor Supplier No. L122812KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137-16-6
Molecular Weight (g/mol) 293.39

Specifications

CAS Max % 1
pH 7.5 to 8.5
Quantity 12 kg
Packaging Poly Bottle
Percent Purity 94%
Grade Ungraded
Chemical Name or Material N to Lauroylsarcosine Sodium Salt

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.