missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Lauroylsarcosine Sodium Salt, 94%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor L1228100GM
| Quantity | 100 g |
|---|---|
| Packaging | Poly Bottle |
Chemical Identifiers
| 137-16-6 | |
| 293.39 |
Specifications
| N to Lauroylsarcosine Sodium Salt | |
| 1 | |
| 100 g | |
| 293.39 | |
| Ungraded |
| 137-16-6 | |
| 7.5 to 8.5 | |
| Poly Bottle | |
| 94% |