Learn More
n-Hexyllithium, 2.5M (33 wt.%) solution in hexane, AcroSeal™, Thermo Scientific Chemicals
$139.50 - $476.39
Chemical Identifiers
| CAS | 21369-64-2 |
|---|---|
| Molecular Formula | C6H13Li |
| Molecular Weight (g/mol) | 92.11 |
| MDL Number | MFCD00191446 |
| InChI Key | CETVQRFGPOGIQJ-UHFFFAOYSA-N |
| Synonym | n-hexyllithium, lithium, hexyl, hexyllithium, hexyl lithium, hexyl-lithium, lithium hexane, n-hexyllithiurn, n-hexyllitium, n-hexyl lithium, hexli |
| PubChem CID | 2733163 |
| IUPAC Name | lithium;hexane |
| SMILES | [Li+].CCCCC[CH2-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC301651000
|
Thermo Scientific Chemicals
301651000 |
100 mL | AcroSeal™ Glass bottle |
Each for $139.50
|
|
||||
|
AC301658000
|
Thermo Scientific Chemicals
301658000 |
800 mL | AcroSeal™ Glass Bottle |
Each for $476.39
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 21369-64-2 | |
| 92.11 | |
| CETVQRFGPOGIQJ-UHFFFAOYSA-N | |
| 2733163 | |
| [Li+].CCCCC[CH2-] |
| C6H13Li | |
| MFCD00191446 | |
| n-hexyllithium, lithium, hexyl, hexyllithium, hexyl lithium, hexyl-lithium, lithium hexane, n-hexyllithiurn, n-hexyllitium, n-hexyl lithium, hexli | |
| lithium;hexane |
Specifications
| 21369-64-2,110-54-3 | |
| 33.2,67.2 | |
| 0.6900g/mL | |
| AcroSeal™ Glass bottle | |
| LiCH2CH2CH2CH2CH2CH3 | |
| 100 mL | |
| n-hexyllithium, lithium, hexyl, hexyllithium, hexyl lithium, hexyl-lithium, lithium hexane, n-hexyllithiurn, n-hexyllitium, n-hexyl lithium, hexli | |
| CETVQRFGPOGIQJ-UHFFFAOYSA-N | |
| lithium;hexane | |
| 2733163 | |
| 32.8 to 33.2% (as HexLi) | |
| n-Hexyllithium, 33 wt. % in n-hexane |
| 32.8,66.8 | |
| Orange to Yellow | |
| −26°C | |
| C6H13Li | |
| MFCD00191446 | |
| 0.69 | |
| Solubility in water: vigorous reaction. | |
| [Li+].CCCCC[CH2-] | |
| 92.11 | |
| 92.11 | |
| Solution |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May cause damage to organs through p
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only