missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyllithium, 2.5M (33 wt.%) solution in hexane, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 301651000

 View more versions of this product

Catalog No. AC301651000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Hexyllithium
21369-64-2,110-54-3
33.2,67.2
0.6900g/mL
AcroSeal™ Glass bottle
LiCH2CH2CH2CH2CH2CH3
100 mL
n-hexyllithium, lithium, hexyl, hexyllithium, hexyl lithium, hexyl-lithium, lithium hexane, n-hexyllithiurn, n-hexyllitium, n-hexyl lithium, hexli
CETVQRFGPOGIQJ-UHFFFAOYSA-N
lithium;hexane
2733163
32.8 to 33.2% (as HexLi)
33 wt. % in n-hexane
32.8,66.8
Orange to Yellow
−26°C
C6H13Li
MFCD00191446
0.69
Solubility in water: vigorous reaction.
[Li+].CCCCC[CH2-]
92.11
92.11
Solution

Chemical Identifiers

21369-64-2
92.11
CETVQRFGPOGIQJ-UHFFFAOYSA-N
2733163
[Li+].CCCCC[CH2-]
C6H13Li
MFCD00191446
n-hexyllithium, lithium, hexyl, hexyllithium, hexyl lithium, hexyl-lithium, lithium hexane, n-hexyllithiurn, n-hexyllitium, n-hexyl lithium, hexli
lithium;hexane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May cause damage to organs through p

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only