accessibility menu, dialog, popup

UserName

CAS RN 21369-64-2

IUPAC Name: lithium(1+) hexan-1-ide
Synonyms: hexyllithium
Molecular Weight (g/mol): 92.11
Molecular Formula: C6H13Li
InChi Key: CETVQRFGPOGIQJ-UHFFFAOYSA-N
SMILES: [Li+].CCCCC[CH2-]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)